2,3-Difluoro-6-methoxybenzoic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:773873-26-0
:
Formula:C₈H₆F₂O₃
MDL Number:MFCD04116018
Molecular Weight:188.13 g/mol
Melting Pt:126…129 °C
EINECS:None
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together