Potassium hydrogen phthalate 99.95-100.05% ACS primary standard
: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
:877-24-7
:
:HOOCC₆H₄COOK
:MFCD00013070
:1.636
:204.22 g/mol
:297 °C (dec.)
:14,7612
:Ambient
:212-889-4
- Environmentally Preferable:
Frequently Bought Together