Potassium hydrogen phthalate 99+%
: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
:877-24-7
:
:HOOCC₆H₄COOK
:MFCD00013070
:204.22 g/mol
:295…300 °C (dec.)
:13,07694
:Ambient
:212-889-4
- Environmentally Preferable:
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration with NaOH:≥99.0 %
- GC:≥97.5 %
Frequently Bought Together