N-Methyl-2-pyrrolidone (NMP) ≥99%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
CAS Number:872-50-4
:
Formula:C₅H₉NO
MDL Number:MFCD00003193
Boiling Pt:202
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Molecular Weight:99.13 g/mol
Flash Pt:91
Merck Index:13,06140
Storage Temperature:Ambient
EINECS:212-828-1
REACH:01-2119472430-46
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99 %
- IR Spectrum:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
spec_VWRC25675.294 spec_VWRC25675.465
Product specification
Frequently Bought Together