Order Entry
Ireland
ContactUsLinkComponent
 
N-Bromosuccinimide for synthesis, Sigma-Aldrich®
 :  Merck
 
N-Bromosuccinimide for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    14.8 g/l (20 °C) (decomposition)
  • Melting Point:
    174 - 179 °C
  • Molar Mass:
    177.98 g/mol
  • Bulk Density:
    750 kg/m3
  • Vapor Pressure:
    14.8 hPa (20 °C)
  • Density:
    2.1 g/cm3 (20 °C)
  • Synonyms:
    NBS
  • Hill Formula:
    C4H4BrNO2

 

Frequently Bought Together