Order Entry
Ireland
ContactUsLinkComponent
 
Dithiothreitol (DTT, Cleland's reagent) 98%, pure
 
undefined
Dithiothreitol (DTT, Cleland's reagent) 98%, pure
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    38 to 43 °C
  • Titration Iodimetric:
    97.5 to 102.5 %

 

 

Frequently Bought Together