Ethyl propionate 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Propionic acid ethyl ester
CAS Number:105-37-3
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00009308
Boiling Pt:99 °C (760 mmHg)
Melting Pt:–73 °C
Density:0.887 g/cm³
Molecular Weight:102.13 g/mol
Flash Pt:12 °C
UN:1195
ADR:3 II
Merck Index:13,03878
EINECS:203-291-4
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3830 to 1.3870 (20°C, 589 nm)
- GC:≥99.0 %
- Relative density:(20/4°C) 0.888 to 0.891
- Acid value:≤1 mg KOH/g
Specification Test Results
Frequently Bought Together