(2S,3S)-(+)-2,3-Butanediol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (2S,3S)-Butane-2,3-diol, (S,S)-(+)-2,3-Butanediol, (2S,3S)-(+)-2,3-Butylene glykol, (S,S)-(+)-2,3-Dihydroxy butane, (2S,3S)-(+)-2,3-Dihydroxy butane, (S,S)-(+)-2,3-Butylene glykol, L(+)-2,3-Butanediol
CAS Number:19132-06-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00063648
Boiling Pt:179…182 °C (760 mmHg)
Density:0.98 g/cm³
Molecular Weight:90.12 g/mol
Flash Pt:85 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4310 to 1.4340 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:+12° to +14° (20°C, 589 nm) (neat)
Specification Test Results
Frequently Bought Together