Platinum, foil, Premion®, Thickness: 1 mm
Catalog # 12060.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:1643
- Environmentally Preferable:
- Pk Info:25×25 mm
Specifications
About this item
Synonyms: Platinium
:
:195.08 g/mol
Boiling Pt:3827
Melting Pt:1769
Density:21.447 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-116-1
CAS Number:7440-06-4
Formula:Pt
MDL Number:MFCD00011179
Merck Index:12,07689