Order Entry
Ireland
ContactUsLinkComponent
Fmoc-Met(O)-OH ≥98% (by HPLC), Novabiochem®
Fmoc-Met(O)-OH ≥98% (by HPLC), Novabiochem®
Certificates
Fmoc-Met(O)-OH ≥98% (by HPLC), Novabiochem®

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity (IR):
    Passes Test
  • Enantiomeric Purity:
    ≥99.0% (a/a)
  • Purity (TLC(011C)):
    ≥98%
  • Purity (TLC(0811)):
    ≥98%
  • Assay (HPLC, area%):
    ≥98.0% (a/a)
  • Solubility (1 mmole in 2 ml DMF):
    Clearly Soluble
  • Assay (Acidimetric):
    ≥95.0%
  • Water (K. F.):
    ≤1.00%

Specification Test Results

Frequently Bought Together