Samarium ≥99.9% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-19-9
:
Formula:Sm
MDL Number:MFCD00011233
Boiling Pt:1900
Melting Pt:1076.85
Density:7.42 g/cm³ (20 °C)
Molecular Weight:150.36 g/mol
Merck Index:12,08498
Storage Temperature:Ambient
EINECS:231-128-7
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together