Order Entry
Ireland
ContactUsLinkComponent
 
Sulphur 99.5+%, refined
 
undefined
Sulphur 99.5+%, refined

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Yellow Powder
  • Assay:
    ≥99.50 %
  • Water:
    ≤0.5 %
  • Sulfated ash:
    ≤0.08 %
  • Acidity:
    ≤0.01 % (as H2SO4)

 

 

Frequently Bought Together