Silicon ≥99.9999% (metals basis), slug, Length: 9.5 mm (0.375 in), Ø 9.5 mm (0.375 in)
Catalog # 43926.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
CAS Number:7440-21-3
:
Formula:Si
MDL Number:MFCD00085311
:28.09 g/mol
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8
Merck Index:12,08635