Phenol : chloroform : iso-amyl alcohol (25:24:1) in saturated aqueous solution, pH 5.2 ready-to-use
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid
CAS Number:136112-00-0
:
MDL Number:MFCD00133763
Density:1.28
Storage Temperature:Refrigerator
UN:2922
ADR:8 II
:GHS06
:GHS05
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together