Order Entry
Ireland
ContactUsLinkComponent
 
Z-AAD-CMK
Catalog # ENZOALX260153R020
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Z-AAD-CMK
Catalog # ENZOALX260153R020
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    20 µl
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Z-AAD-CMK
  • Purity:
    >95% (HPLC)
  • Concentration:
    10 mM
  • Formulation:
    Liquid. In DMSO.

Specifications

About this item

Granzyme B inhibitor

Synthetic peptide that irreversibly inhibits granzyme B activity. The inhibitor is desigend as a methyl ester to facilitate cell permeability.