Order Entry
Ireland
ContactUsLinkComponent
Suc-Leu-Leu-Val-Tyr-AMC
Suc-Leu-Leu-Val-Tyr-AMC
Catalog # ENZOBMLP8020005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Suc-Leu-Leu-Val-Tyr-AMC
Catalog # ENZOBMLP8020005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Source:
  • Species:
  • Seq:
    02
  • Environmentally Preferable:
  • Protein/peptide name:
    Suc-Leu-Leu-Val-Tyr-AMC
  • Purity:
    >99% (TLC), peptide content: 98%
  • Formulation:
    Lyophilised

Specifications

About this item

Proteasome substrate

Fluorogenic substrate for 20S proteasome (Prod. No. BML-PW8720) and other chymotrypsin-like proteases, as well as calpains. Ex.: 380 nm Em.: 460 nm.