Cell counting kit-8
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages: One-step, ready-to-use solution, no radioisotopes, no solubilization step, high sensitivity, correlates with the [3H]-thymidine incorporation assay. Can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results.
Ordering information: Quantity: 500 tests/5ml; 2500 tests/5 x 5ml. Contents: 5mM WST-8 solution, 0.2mM 1-methoxy-PMS, 150mM sodium chloride.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together