Order Entry
Ireland
ContactUsLinkComponent
Anti-GCDFP15 Mouse Monoclonal Antibody
Anti-GCDFP15 Mouse Monoclonal Antibody
Catalog # ENZOALX801051L001
Supplier:  ENZO LIFE SCIENCES
New Product
Anti-GCDFP15 Mouse Monoclonal Antibody
Catalog # ENZOALX801051L001
Supplier:  ENZO LIFE SCIENCES
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Gross Cystic Disease Fluid Protein 15
  • Antigen symbol:
    GCDFP15
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
  • Seq:
    02
  • Environmentally Preferable:
  • Form:
    Liquid. In 0,01 M PBS containing 1% BSA and 0,1% sodium azide .
  • Antigen synonyms:
    GCDGP-15|SABP|Gross cystic disease fluid protein 15|Gp17|GPIP4|Prolactin induced protein|Prolactin inducible protein precursor|Secretory actin binding protein.|GCDGP 15
  • Pk:
    1 mL

Specifications

About this item

Gross cystic disease is a common premenopausal disorder in which gross cysts are the predominant pathologic lesion. It is characterized by production of a fluid secretion which accumulates in the breast cysts. Gross cystic disease fluid is a pathologic secretion from breast composed of several glycoproteins, including a unique 15kDa monomer protein, GCDFP-15. The cells within the body that produce GCDFP-15 appear to be restricted primarily to those with apocrine function. Studies have found GCDFP-15 to be a highly specific and sensitive marker for breast cancer.
GCDFP-15 is expressed by apocrine sweat glands, eccrine glands (variable), minor salivary glands, bronchial glands, metaplasmic epithelium of the breast, benign sweat gland tumors of the skin and the serous cells of the submandibular gland. Breast carcinomas (primary and metastatic lesions) with apocrine features express GCDFP-15. GCDFP-15 is positive in extramammary Paget's Disease. Other tumors tested are negative.

  • For Immunohistochemistry

Recognizes human GCDFP-15.

Type: Primary
Antigen: GCDFP-15
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human