Anti-P2X7 Rabbit Polyclonal Antibody
Catalog # ENZOALX215035R100
Supplier: ENZO LIFE SCIENCES
New Product
Specifications
- Antibody type:Primary
- Antigen name:Purinergic Receptor P2X, Ligand-gated Ion channel 7
- Antigen symbol:P2X7
- Clonality:Polyclonal
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Mouse
- Seq:02
- Environmentally Preferable:
- Form:Liquid. Antiserum diluted with PBS containing 0,1% BSA and 0,02% sodium azide.
- Pk:100 µl
Specifications
About this item
Host: Rabbit
- For Immunocytochemistry, Immunoprecipitation
Recognizes mouse purinergic receptor P2X7.
Type: Primary
Antigen: P2X7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse