Anti-NFkB p105/P50 Polyclonal Antibody
BSBTPB9149
- Antibody type:Primary
- Antigen symbol:NFkB p105/P50
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for P50/NFKB1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. P50/NFKB1 information: Molecular Weight: 105356 MW; Subcellular Localization: Nucleus. Cytoplasm. Nuclear, but also found in the cytoplasm in an inactive form complexed to an inhibitor (I-kappa-B).
Type: Primary
Antigen: NFkB p105/P50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat