Anti-Collagen III Polyclonal Antibody
Catalog # BSBTRP1033
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Collagen III
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for COLLAGEN III/COL3A1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. COLLAGEN III/COL3A1 information: Molecular Weight: 138564 MW; Subcellular Localization: Secreted, extracellular space, extracellular matrix .
Type: Primary
Antigen: Collagen III
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human