Anti-PPBP Mouse Monoclonal Antibody [clone: G6_5C7.3]
Catalog # PRSIXP-5717-M
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Pro-Platelet Basic Protein
- Antigen symbol:PPBP
- Clonality:Monoclonal
- Clone:G6_5C7.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:5473
- Antigen synonyms:Beta-TG|CTAPIII|SCYB7|CTAP3|TC2|TGB|CTAP-III|MDGF|LDGF|PBP|THBGB1|LA-PF4|THBGB|TC1|TGB1|NAP-2|CXCL7|B-TG1
- Storage temperature:Stable at room temperature for at least one month and for greater than a year when kept at –20 °C. Reconstitute in sterile water to a concentration of 0.1 - 1.0 mg/mL. Stable for at least six weeks at 2-4 °C when reconstituted.
- Shipping temperature:RT
- Immunogen:The monoclonal antibody was produced in BALB/c mice using recombinant human NAP-2 as the immunizing antigen.
- Purification:This IgG1/K antibody was purified from a Protein A chromatography column.
- Size:500 µg
- Pk:500 µG
Specifications
About this item
NAP2 is a platelet-derived growth factor that belongs to the CXC chemokine family. This growth factor is a potent chemoattractant and activator of neutrophils. It has been shown to stimulate various cellular processes including DNA synthesis, mitosis, glycolysis, intracellular cAMP accumulation, prostaglandin E2 secretion, and sythesis of hyaluronic acid and sulfated glycosaminoglycan. It also stimulates the formation and secretion of plasminogen activator by synovial cells.
Type: Primary
Antigen: PPBP
Clonality: Monoclonal
Clone: G6_5C7.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human