Specifications
- Antigen name:Polymerase I and Transcript Release Factor
- Antigen symbol:PTRF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:CAVIN|cavin-1|FKSG13|Polymerase I and transcript release factor|CAVIN1|PTRF|Cavin-1|CGL4
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal PTRF antibody was raised against a 19 amino acid peptide near the carboxy terminus of human PTRF.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PTRF Antibody: The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3' end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
PTRF Antibody is affinity chromatography purified via peptide column.
PTRF Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PTRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human