Anti-WLS Chicken Polyclonal Antibody
Catalog # PRSI54-212
Supplier: ProSci Inc.
Specifications
- Antigen symbol:WLS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Immunogen:Human GPR177 C-terminus peptide
- Size:100 µl
- Pk:100 µl
Specifications
About this item
GPR177 antibody can be used at 1:2,000 to 1:5,000 for immunoblot testing. User needs to optimize the concentrations in assay.
Affinity Purified
PBS Buffer containing 0.02% sodium azide.
2-8°C for one week and -20°C as an undiluted liquid for up to two years.
Type:
Antigen: WLS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Bovine, Human, Mouse, Rat