- Antigen name:Jagged 1
- Antigen symbol:JAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Ser-1|Htu|AWS|AHD|Jag1|jagged|JAGL1|ABE2|HJ1|Ozz|AGS|Gsfabe2|CD339
- Immunogen:JAG1 antibody was raised against a synthetic peptide corresponding to aa 110 - 125 of JAG1 (Human).
- Size:50 µg
- Pk:50 µG
Jagged-1 (JAG1) is a cell surface protein which is one of several ligands that activate Notch and related receptors. JAG1 functions in an embryologically important signaling pathway and has also been shown to play a role in hematopoiesis. JAG1 inhibits proliferation of cd34+ macrophage progenitor cells and a JAG1 gene abnormality may be an aggravating factor in extrahepatic biliary atresia. Activation of NOTCH1 signaling by JAG1 induces monocyte-derived dendritic cell maturation in vitro. JAG1 interaction with NOTCH1 on tumor cells dramatically induces proliferation and inhibition of apoptosis in vitro. Either haploinsufficiency for wild-type JAG1 and/or dominant negative effects produced by mutated JAG1 are responsible for the Alagille syndrome. A BLAST analysis was used to suggest crossreactivity with Jagged-1 protein from human, chimpanzee, rat and mouse based on 100% homology with the immunizing sequence. Partial reactivity is expected against canine (81%) and Xenopus laevis (85%) based on partial sequence homologies as indicated. The jagged 1 protein encoded by JAG1 is the human homolog of the Drosophilia jagged protein. Human jagged 1 is the ligand for the receptor notch 1, the latter a human homolog of the Drosophilia jagged receptor notch. Mutations that alter the jagged 1 protein cause Alagille syndrome. Jagged 1 signalling through notch 1 has also been shown to play a role in hematopoiesis.
JAG1 antibody can be used in Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunocytochemistry.
Protein A Column
PBS, pH 7.2, 0.01% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: JAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human