Anti-CCL3L1 Goat Polyclonal Antibody
PRSI46-969
- Antigen name:Chemokine (C-C motif) ligand 3-like 1
- Antigen symbol:CCL3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:CCL3
- Concentration:500 ug/mL
- Immunogen:CCL3L1 antibody was raised against a 15 amino acid synthetic peptide near the internal region of CCL3L1.
- Size:100 µg
- Pk:100 µG
CCL3L1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
CCL3L1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CCL3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: