Order Entry
Ireland
ContactUsLinkComponent
Anti-LARP1B Goat Polyclonal Antibody
Anti-LARP1B Goat Polyclonal Antibody
Catalog # PRSI45-821
Supplier:  ProSci Inc.
undefined
Anti-LARP1B Goat Polyclonal Antibody
Catalog # PRSI45-821
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    La ribonucleoprotein Domain Family member 1B
  • Antigen symbol:
    LARP1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    LARP2
  • Concentration:
    500 ug/mL
  • Immunogen:
    LARP2 antibody was raised against a 14 amino acid synthetic peptide near the internal region of LARP2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

LARP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LARP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LARP1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human