Order Entry
Ireland
ContactUsLinkComponent
Anti-CABP1 Goat Polyclonal Antibody
Anti-CABP1 Goat Polyclonal Antibody
Catalog # PRSI45-349
Supplier:  ProSci Inc.
undefined
Anti-CABP1 Goat Polyclonal Antibody
Catalog # PRSI45-349
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Calcium Binding Protein 1
  • Antigen symbol:
    CABP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    Calbrain|CABP1|CaBP1
  • Concentration:
    500 ug/mL
  • Immunogen:
    CABP1 antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of CABP1.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

CABP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CABP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: 0
Antigen: CABP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human