Anti-ADRB3 Goat Polyclonal Antibody
Catalog # PRSI42-948
Supplier: ProSci Inc.
Specifications
- Antigen name:Adrenoceptor Beta 3
- Antigen symbol:ADRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Adrb-3|FLJ99960|BETA3AR
- Concentration:500 ug/mL
- Immunogen:Adrb3 Antibody was raised against a 12 amino acid sequence near the internal region (near C-Terminus) of Adrb3 (mouse, intracellular).
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Adrb3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: