Anti-ADAMTS8 Goat Polyclonal Antibody
PRSI42-545
- Antigen name:ADAM Metallopeptidase with Thrombospondin type 1 motif, 8
- Antigen symbol:ADAMTS8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:ADAMTS8
- Concentration:500 ug/mL
- Immunogen:ADAMTS8 antibody was raised against a 10 amino acid synthetic peptide near the internal region of ADAMTS8 (aa 176-185).
- Size:100 µg
- Pk:100 µG
ADAMTS8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ADAMTS8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADAMTS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: