Anti-NOV Rabbit Polyclonal Antibody (Biotin)
Catalog # PRSI38-186
Supplier: ProSci Inc.
Specifications
- Antigen name:Nephroblastoma overexpressed
- Antigen symbol:NOV
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:IBP-9|IGFBP-9|NOVh|IGFBP9|CCN3
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hNOV. Human NOV specific antibody was purified by affinity chromatography and then biotinylated.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: NOV
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human