Anti-SERPINH1 Rabbit Polyclonal Antibody
PRSI31-063
- Antigen name:Serpin Family H Member 1
- Antigen symbol:SERPINH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Serpin H1|Cell proliferation-inducing gene 14 protein|CBP1|Arsenic-transactivated protein 3|HSP47|AsTP3|47 kDa heat shock protein|Rheumatoid arthritis-related antigen RA-A47|SERPINH2|Collagen-binding protein|PIG14|SERPINH1|CBP2|Colligin
- Immunogen:A synthetic peptide corresponding a region of human SERPINH1.
- Size:100 µg
- Pk:100 µG
Serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
Antibody is purified by Protein A/G affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SERPINH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human