Anti-PWP1 Rabbit Polyclonal Antibody
Catalog # PRSI30-633
Supplier: ProSci Inc.
Specifications
- Antigen name:PWP1 homolog (S. cerevisiae)
- Antigen symbol:PWP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human PWP1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
PWP1 belongs to the WD repeat PWP1 family. It contains 5 WD repeats. PWP1 may play an important role in cell growth and/or transcription.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PWP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat