Anti-NR5A1 Rabbit Polyclonal Antibody
PRSI30-083
- Antigen name:Nuclear Receptor Subfamily 5 Group A Member 1
- Antigen symbol:NR5A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:SF-1|AD4BP|FTZF1|FTZ1|SF1|ELP|POF7
- Immunogen:A synthetic peptide corresponding a region of human NR5A1.
- Size:50 µg
- Pk:50 µG
NR5A1 is an important regulator of steroidogeneisis which is present in human skin and its appendages. It plays a role in regulating p450scc expression with TReP-132 and CBP/p300.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NR5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human