Anti-MANSC1 Rabbit Polyclonal Antibody
Catalog # PRSI28-062
Supplier: ProSci Inc.
Specifications
- Antigen name:MANSC Domain Containing 1
- Antigen symbol:MANSC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Loss Of Heterozygosity 12 Chromosomal Region 3 Protein|LOH12CR3|MANSC Domain Containing 1
- Immunogen:A synthetic peptide corresponding a region of human MANSC1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
MANSC1, located on chromosome 12, encodes a protein whose function is not yet defined.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: MANSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human