Anti-RUVBL2 Rabbit Polyclonal Antibody
Catalog # PRSI27-571
Supplier: ProSci Inc.
Specifications
- Antigen name:RuvB-like 2
- Antigen symbol:RUVBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:REPTIN|RVB2|TAP54-beta|INO80J|ECP-51|RuvB like AAA ATPase 2|TIP48|TIH2|CGI-46|TIP49B|ECP51
- Immunogen:A synthetic peptide corresponding a region of human RUVBL2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
RUVBL2 encodes the second human homologue of the bacterial RuvB gene. Bacterial RuvB protein is a DNA helicase essential for homologous recombination and DNA double-strand break repair. Functional analysis showed that this gene product has both ATPase and DNA helicase activities.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RUVBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat