Anti-JMJD8 Rabbit Polyclonal Antibody
Catalog # PRSI27-461
Supplier: ProSci Inc.
Specifications
- Antigen name:Jumonji Domain-Containing protein 8
- Antigen symbol:JMJD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Jumonji domain-containing protein 8|JMJD8|PP14397|C16orf20|JmjC domain-containing protein 8
- Immunogen:A synthetic peptide corresponding a region of human LOC339123.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
LOC339123 is a candidate transcription factor located on chromosome 16.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: JMJD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human