Anti-TXNDC5 Rabbit Polyclonal Antibody
Catalog # PRSI27-186
Supplier: ProSci Inc.
Specifications
- Antigen name:Thioredoxin Domain Containing 5
- Antigen symbol:TXNDC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:PDIA15|UNQ364|HCC2|STRF8|ERP46|HCC-2|ENDOPDI
- Immunogen:A synthetic peptide corresponding a region of human TXNDC5.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
TXNDC5 is a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis.This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. This gene can be co-transcribed with the upstream gene MU.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TXNDC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human