Order Entry
Ireland
ContactUsLinkComponent
 
Anti-STK16 Rabbit Polyclonal Antibody
  PRSI26-300
undefined
Anti-STK16 Rabbit Polyclonal Antibody
  PRSI26-300

 

  • Antigen name:
    Serine/Threonine Kinase 16
  • Antigen symbol:
    STK16
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Antigen synonyms:
    hPSK|Myristoylated and palmitoylated serine-threonine kinase|Protein kinase PKL12|TSF-1|MPSK|TGF-beta stimulated factor 1|Serine/threonine protein kinase 16
  • Immunogen:
    A synthetic peptide corresponding a region of human STK16.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

STK16 is a protein kinase that acts on both serine and threonine residues.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STK16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human