Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ADAM9 Rabbit Polyclonal Antibody
Catalog # PRSI26-131
Supplier:  ProSci Inc.
undefined
Anti-ADAM9 Rabbit Polyclonal Antibody
Catalog # PRSI26-131
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ADAM Metallopeptidase Domain 9
  • Antigen symbol:
    ADAM9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Antigen synonyms:
    ADAM9|KIAA0021|Metalloprotease/disintegrin/cysteine-rich protein 9|Meltrin-gamma|MLTNG|Cellular disintegrin-related protein|A disintegrin and metalloproteinase domain 9|ADAM 9|Myeloma cell metalloproteinase|MCMP|MDC9
  • Immunogen:
    A synthetic peptide corresponding a region of human ADAM9.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

ADAM9 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. ADAM9 interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor.This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor. Two alternative splice variants have been identified, encoding distinct isoforms.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADAM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat