- Antigen name:Gamma-glutamyl Carboxylase
- Antigen symbol:GGCX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:VKCFD1
- Immunogen:A synthetic peptide corresponding a region of human GGCX.
- Size:50 µg
- Pk:50 µG
GGCX is an enzyme which catalyzes the posttranslational modification of vitamin K-dependent protein. Many of these vitamin K-dependent proteins are involved in coagulation so the function of the encoded enzyme is essential for hemostasis. Mutations in this gene are associated with vitamin K-dependent coagulation defect and PXE-like disorder with multiple coagulation factor deficiency.Gamma-glutamyl carboxylase accomplishes the posttranslational modification required for the activity of all of the vitamin K-dependent proteins (Wu et al., 1991 [PubMed 1749935]). These include some of the blood coagulation and anticoagulation proteins as well as bone gamma-carboxyglutamic acid (Gla) protein (BGLAP; MIM 112260) and bone matrix protein (MGP; MIM 154870).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GGCX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat