Specifications
- Antigen name:Epithelial Splicing Regulatory Protein 2
- Antigen symbol:ESRP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:RBM35B|RNA Binding Motif Protein 35B
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:RBM35B antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human RBM35B.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RBM35B Antibody: RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5' UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing programme induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.
Type:
Antigen: ESRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse