- Antigen name:Thyroid Hormone Receptor Interactor 6
- Antigen symbol:TRIP6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:MGC10558|MGC10556|MGC29959|zyxin related protein 1|thyroid receptor-interacting protein 6|TRIP6|MGC4423|MGC3837|OIP1|thyroid hormone receptor interactor 6|ZRP-1|OPA-interacting protein 1
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:TRIP6 antibody was raised against a 15 amino acid synthetic peptide near the center of human TRIP6.
- Tested applications:E, WB, ICC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
TRIP6 Antibody: TRIP6 (Thyroid receptor interacting protein 6) is a zyxin-related focal adhesion molecule involved in cell motility and antiapoptotic responses. TRIP6 is predominantly expressed in kidney, liver and lung. It has three LIM domains, a PDZ-binding motif, a Crk SH2-binding motif and/or other protein interacting domains. TRIP6 regulates NF-kappa B activation and interacts with RICK and TRAF2, proteins that are involved in TNF signaling. TRIP6 antagonizes Fas-induced apoptosis and promotes Fas-mediated cell migration in apoptosis-resistant glioma cells via Src-dependent phosphorylation of TRIP6 at Tyr-55.
Type:
Antigen: TRIP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse