Order Entry
Ireland
ContactUsLinkComponent
Anti-HVCN1 Rabbit Polyclonal Antibody
Anti-HVCN1 Rabbit Polyclonal Antibody
Catalog # PRSI5875
Supplier:  ProSci Inc.
undefined
Anti-HVCN1 Rabbit Polyclonal Antibody
Catalog # PRSI5875
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Hydrogen Voltage-Gated Channel 1
  • Antigen symbol:
    HVCN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Antigen synonyms:
    Hydrogen voltage-gated channel 1|HVCN1|HV1|Voltage-gated hydrogen channel 1|VSOP|UNQ578/PRO1140
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    HVCN1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human HVCN1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

HVCN1 Antibody: HVCN1 (hydrogen voltage-gated channel 1), a voltage gated proton channel protein that is highly expressed in immune tissues, mediates the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing. HVCN1 moderates the voltage-dependent proton permeability of excitable membranes by allowing the flow of protons in accordance to their electrochemical gradient. HVCN1 is sensitive to zinc ions, and can be inhibited by them. HVCN1 may represent a new therapeutic target for B cell malignancies on continued signaling via the B cell antigen receptor.

Type:
Antigen: HVCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat