Specifications
- Antigen name:Duffy antigen/chemokine receptor
- Antigen symbol:DARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:ACKR1|Atypical chemokine receptor 1|CCBP1|CD234|GpFy|GPD|WBCQ1|FY|Dfy
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:DARC antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human DARC.
- Tested applications:E, WB, IHC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DARC Antibody: DARC, also known as the Duffy antigen/chemokine receptor, is a seven-transmembrane protein homologous to the classical chemokine G-protein coupled receptors (GPCRs) with the exception of the motif required for G protein coupling. DARC can bind with high affinity several chemokines without transducing any signal, suggesting it may modulate the signals normally induced by these chemokines. Recently, DARC was found to interact with KAI1, a four transmembrane protein recently identified as a tumor metastasis suppressor protein. It is thought that tumor cells dislodged from the primary tumor and expressing KAI1 interact with DARC proteins expressed on vascular cells, transmitting a senescent signal to the tumor cells, while tumor cells that have lost KAI1 expression can proliferate and potentially give rise to metastases. At least three isoforms of DARC are known to exist.
Type:
Antigen: DARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human