Lock rails for EZ panel Loc™
Supplier: Brady
Certificates
About this item
The unique lock rail system allows panel door to be closed even when lockouts are in place and accommodates multiple locks, eliminating the need for hasps during group lockouts.
- Lock rail design keeps weight off the breakers which protects delicate electrical components from damage
- Lock rails made of durable Isoplast polymers with acrylic adhesive backing
- 'No Shackle' locks with EZ Panel Loc keys are sold separately and are especially useful when you want lockout devices to be kept at the point of need at the panel
Frequently Bought Together