Containers, stainless steel
These containers are made of 316L stainless. The high quality crevice-free finish makes these containers ideal for powders, liquids and most demanding applications.
- Multipurpose containers
- Robust construction
- All welds ground and polished
- Standard 102 or 51 mm lid (Tri-ferrule style)
- Complete with lid, silicone gasket and clamp band
The handy 101,6 mm tri clamp connection on the top of the containers can be used to clamp on to existing similar equipment.
Frequently Bought Together