Anti-Claudin-3 Rabbit Polyclonal Antibody
BIRBORB160459-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Claudin-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Storage temperature:Store at –20 °C for a year
- Immunogen:Synthesized peptide derived from the C-terminal region of human Claudin-3.
- Pk:100 µl
Type: Primary
Antigen: Claudin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat