Seal with Bore for Screw Caps, Silicone with PTFE Washer
Silicone rubber sealing rings with PTFE washers.
- Temperature resistant from −200 to +250 °C
- Excellent chemical resistance
- Easy handling
For threaded connection caps with central bore and GL thread.
Frequently Bought Together