Order Entry
Ireland
ContactUsLinkComponent
Neutralizing agent, RBS NA 2
Neutralizing agent, RBS NA 2
Catalog # CHEPRBSNA2
Supplier:  CHEMICAL
undefined
Neutralizing agent, RBS NA 2
Catalog # CHEPRBSNA2
Supplier:  CHEMICAL
Specifications
  • Description:
    Neutralizing agent, RBS NA 2
  • Environmentally Preferable:
  • Pk:
    5 L
  • pH range:
    1.8 (0.5%)
Specifications
About this item
Improves the cleaning results in washing machines - labwashers.
  • Concentrated solution
  • Surfactant-free
  • Based on phosphoric acid
RBS NA 2 solution allows neutralisation and removal of residual alkaline traces and mineral salts from the cleaned items and the internal components of the washing machine. Can also be used in the pre-wash cycle in case of heavy residues.
Ordering information: Available in box of 4 x 5 litres.